Trinatriumtrimetaphosphat

Lieferant: Thermo Scientific

Synonyme: Trisodium trimetaphosphate

89063.A1 89063.A7
89063.A1EA 67.1 EUR
89063.A1 89063.A7
Trinatriumtrimetaphosphat
Trinatriumtrimetaphosphat

CAS No.: 7785-84-4 Molecular Formula: (NaPO3)3 Formula Weight: 305.89 Physical Form: Powder Density: 2.49

Formel: Na₃O₉P₃
Molecular Weight: 305,89 g/mol
Schmelzpunkt: 588 °C
Dichte: 2,49
Lagertemperatur: Raumtemperatur
MDL: MFCD00867826
CAS-Nummer: 7785-84-4
EINECS: 232-088-3
Merck Index: 14,8696

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR