tri-Kaliumorthophosphat, wasserfrei 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Kaliumphosphat, Kaliumphosphat tribasisch
41587.30
41587.A1
41587.A7
41587.30EA
95.7
EUR
41587.30
41587.A1
41587.A7
tri-Kaliumorthophosphat, wasserfrei 97%
tri-Kaliumorthophosphat
Purity: 97%
CAS No.: 7778-53-2
Molecular Formula: K3PO4
Formula Weight: 212.27
Physical Form: Granular
Density: 2.564
Melting Point: 1340°
Solubility: Soluble in water. Insoluble in alcohol
Merck Reference: 14,7660
MDL No.: MFCD00036295
Formel:
K₃O₄P Molecular Weight: 212.27 g/mol Schmelzpunkt: 1340 Dichte: 2.564 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036295 CAS-Nummer: 7778-53-2 EINECS: 231-907-1 Merck Index: 14,7660 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...