Bariumcarbonat 99+% zur Analyse

Lieferant: Thermo Fisher Scientific
Achtung

217495000 217490025
ACRO217495000EA 52.9 EUR
ACRO217495000 ACRO217490025
Bariumcarbonat 99+% zur Analyse
Barium carbonate
Formel: BaCO₃
Molecular Weight: 197.34 g/mol
Siedepunkt: 1450 °C
Schmelzpunkt: 811 °C
Dichte: 4.43 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00003448
CAS-Nummer: 513-77-9
EINECS: 208-167-3
Merck Index: 13,00972

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Titration Complexometric ≥99.0 %
Chloride (Cl) ≤300 ppm
Sulfide (S) ≤10 ppm
Calcium (Ca) ≤1000 ppm
Iron (Fe) ≤50 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤2000 ppm
Strontium (Sr) ≤10000 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.015 % (in dilute HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR