Bariumcarbonat 99+% ACS

Lieferant: Thermo Scientific
Achtung

ACRO423440025EA 158 EUR
ACRO423440025 ACRO423445000
Bariumcarbonat 99+% ACS
Barium carbonate
Formel: BaCO₃
Molecular Weight: 197,34 g/mol
Siedepunkt: 1450 °C
Schmelzpunkt: 811 °C
Dichte: 4,43 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00003448
CAS-Nummer: 513-77-9
EINECS: 208-167-3
ADR: 6.1,III
Merck Index: 13,00972

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Titration excess HCl 99.0 to 101.0 %
Chloride (Cl) ≤0.002 %
Sulfide (S) ≤0.001 %
Calcium (Ca) ≤0.05 %
Iron (Fe) ≤0.002 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.02 %
Strontium (Sr) ≤0.7 %
Heavy metals (as Pb) ≤10 ppm
Water soluble titr. base ≤0.002 meq/g
Insoluble matter ≤0.015 % (in dilute HCl)
Oxidizing substance (as NO3) ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR