Bariumcarbonat ≥99,95% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

14452.18 14452.30 14452.A1 14452.09
14452.18EA 87.7 EUR
14452.18 14452.30 14452.A1 14452.09
Bariumcarbonat ≥99,95% (Metall-Basis)
Barium carbonate

Purity: 99.95% CAS No.: 513-77-9 Molecular Formula: BaCO3 Formula Weight: 197.35 Physical Form: Lump/Powder Density: 4.43 Melting Point: 811° Boiling Point: dec. Merck Reference: 14,969 MDL No.: MFCD00003448

Formel: BaCO₃
Molecular Weight: 197.34 g/mol
Siedepunkt: Zersetzung
Schmelzpunkt: 811 °C
Dichte: 4.43
Lagertemperatur: Raumtemperatur
MDL: MFCD00003448
CAS-Nummer: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1 III
Merck Index: 14,969

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR