Bariumcarbonat, tech.

Lieferant: Thermo Scientific
Achtung

43478.22 43478.A9 43478.A3 43478.36
43478.22EA 16.7 EUR
43478.22 43478.A9 43478.A3 43478.36
Bariumcarbonat, tech.
Barium carbonate

CAS No.: 513-77-9 Molecular Formula: BaCO3 Formula Weight: 197.35 Physical Form: Powder Density: 4.43 Odour: Odourless Boiling Point: dec. Solubility: Almost insoluble in water. Soluble in NH4Cl solution, dilute HCl, HNO3, acetic acid. Insoluble in most alkalies Merck Reference: 14,969 MDL No.: MFCD00003448


Used in the manufacture of glass, brick, clay products, barium chemicals, barium ferrites, and production of photographic papers

Formel: BaCO₃
Molecular Weight: 197,34 g/mol
Siedepunkt: Zersetzung
Schmelzpunkt: 811 °C
Dichte: 4,43
Lagertemperatur: Raumtemperatur
MDL: MFCD00003448
CAS-Nummer: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1,III
Merck Index: 14,969

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR