Calciumcarbonat ≥99,99% (Metall-Basis), Puratronic®
Lieferant: Thermo Scientific
Synonyme:
Marmor
10679.14
10679.22
10679.36
10679.06
10679.14EA
238
EUR
10679.14
10679.22
10679.36
10679.06
Calciumcarbonat ≥99,99% (Metall-Basis), Puratronic®
Calciumcarbonat
Purity: 99.99%
CAS No.: 471-34-1
Molecular Formula: CaCO3
Formula Weight: 100.09
Physical Form: Powder
Density: 2.930
Refractive Index: 1.6583
Appearance: White
Odour: Odourless
Melting Point: 800° dec.
Merck Reference: 14,1657
MDL No.: MFCD00010906
Formel:
CaCO₃ Molecular Weight: 100,09 g/mol Schmelzpunkt: 800 °C (Zers.) Dichte: 2,930 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010906 CAS-Nummer: 471-34-1 EINECS: 207-439-9 Merck Index: 14,1657 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...