Calciumcarbonat ≥99,997% (Metall-Basis), Puratronic®
Lieferant: Thermo Scientific
Synonyme:
Marmor
43073.22
43073.14
43073.36
43073.06
43073.22EA
818
EUR
43073.22
43073.14
43073.36
43073.06
Calciumcarbonat ≥99,997% (Metall-Basis), Puratronic®
Calciumcarbonat
Purity: 99.997%
CAS No.: 471-34-1
Molecular Formula: CaCO3
Formula Weight: 100.09
Physical Form: Powder
Density: 2.930
Refractive Index: 1.6583
Appearance: White
Odour: Odourless
Melting Point: 800° dec.
Merck Reference: 14,1657
MDL No.: MFCD00010906
Formel:
CaCO₃ Molecular Weight: 100,09 g/mol Schmelzpunkt: 800 °C (Zers.) Dichte: 2,930 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010906 CAS-Nummer: 471-34-1 EINECS: 207-439-9 Merck Index: 14,1657 |