Strontiumchromat ≥95%

Lieferant: Thermo Scientific
Gefahr

89026.30 89026.A1
89026.30EA 81.3 EUR
89026.30 89026.A1
Strontiumchromat ≥95%
Strontium chromate

CAS No.: 7789-06-2 Molecular Formula: SrCrO4 Formula Weight: 203.61 Physical Form: Powder Density: 3.895 Odour: Odourless Merck Reference: 14,8841 UN No.: 3087 MDL No.: MFCD00049549

Formel: CrO₄Sr
Molecular Weight: 203,61 g/mol
Dichte: 3,895
Lagertemperatur: Raumtemperatur
MDL: MFCD00049549
CAS-Nummer: 7789-06-2
EINECS: 232-142-6
UN: 3087
ADR: 5.1,III
Merck Index: 14,8841

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR