Bariumfluorid, ultra trocken ≥99,998% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung


Unter Argon in Ampullen verpackt

38610.09 38610.18
38610.09EA 99.4 EUR
38610.09 38610.18
Bariumfluorid, ultra trocken ≥99,998% (Metall-Basis)
Barium fluoride

Purity: 99.998% CAS No.: 7787-32-8 Molecular Formula: BaF2 Formula Weight: 175.36 Physical Form: Powder, Ampouled under argon Density: 4.83 Appearance: White Odour: Odourless Melting Point: 1353° Boiling Point: 2260° Merck Reference: 14,974 UN No.: 1564 MDL No.: MFCD00003450

Formel: BaF₂
Molecular Weight: 175,33 g/mol
Siedepunkt: 2260 °C
Schmelzpunkt: 1353 °C
Dichte: 4,83
Lagertemperatur: Raumtemperatur
MDL: MFCD00003450
CAS-Nummer: 7787-32-8
EINECS: 232-108-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,974

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR