Magnesiumfluorid ≥99% (Metall-Basis ausgeschlossen Ca, Na) (max. 1% Ca + Na)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99%
CAS No.: 7783-40-6
Molecular Formula: MgF2
Formula Weight: 62.30
Physical Form: -200 Mesh Powder
Density: 3.148
Refractive Index: 1.35-1.38
Appearance: White to off-white
Odour: Odourless
Melting Point: 1261°
Solubility: Sparingly soluble in water. Slightly soluble in dilute HNO3
Merck Reference: 14,5665
MDL No.: MFCD00011108
Formel:
MgF₂ Siedepunkt: 2260 °C (1013 hPa) Schmelzpunkt: 1261 °C Dichte: 3,148 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011108 CAS-Nummer: 7783-40-6 EINECS: 231-995-1 Merck Index: 14,5665 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...