Strontiumfluorid ≥99,99% (Metall-Basis), Puratronic®
Lieferant: Thermo Scientific
|
Achtung
|
10878.06
10878.14
10878.22
10878.06EA
65.7
EUR
10878.06
10878.14
10878.22
Strontiumfluorid ≥99,99% (Metall-Basis), Puratronic®
Strontium fluoride
Purity: 99.99%
CAS No.: 7783-48-4
Molecular Formula: SrF2
Formula Weight: 125.62
Physical Form: Powder
Density: 4.240
Refractive Index: 1.442
Appearance: White
Odour: Odourless
Melting Point: 1473°
Merck Reference: 14,8842
MDL No.: MFCD00011251
Formel:
F₂Sr Molecular Weight: 125,62 g/mol Siedepunkt: 2460 °C (1013 hPa) Schmelzpunkt: 1473 °C Dichte: 4,240 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011251 CAS-Nummer: 7783-48-4 EINECS: 232-000-3 UN: 3288 ADR: 6.1,III Merck Index: 14,8842 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...