Strontiumfluorid ≥99%

Lieferant: Thermo Scientific
Achtung

12201.30 12201.A1 12201.A7
12201.30EA 76.5 EUR
12201.30 12201.A1 12201.A7
Strontiumfluorid ≥99%
Strontium fluoride

Purity: 99% min CAS No.: 7783-48-4 Molecular Formula: SrF2 Formula Weight: 125.62 Physical Form: Powder Density: 4.240 Refractive Index: 1.442 Appearance: White Odour: Odourless Melting Point: 1473° Solubility: Slightly soluble in water (11.7mg at 18°C/100ml), dilute acids. Decomposes in strong acids Merck Reference: 14,8842 MDL No.: MFCD00011251

Formel: F₂Sr
Molecular Weight: 125,62 g/mol
Siedepunkt: 2460 °C (1013 hPa)
Schmelzpunkt: 1473 °C
Dichte: 4,240
Lagertemperatur: Raumtemperatur
MDL: MFCD00011251
CAS-Nummer: 7783-48-4
EINECS: 232-000-3
UN: 3288
ADR: 6.1,III
Merck Index: 14,8842

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR