Calciumoxid, Reagenzqualität
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: Quicklime, Calcium oxide from marble, caustic, Lime
33299.36 33299.A3
33299.36EA 76.2 EUR
33299.36 33299.A3
Calciumoxid, Reagenzqualität
Calcium oxide
CAS No.: 1305-78-8 Molecular Formula: CaO Formula Weight: 56.08 Physical Form: Powder Density: 3.37 Refractive Index: 1.83 Sensitivity: Air and Moisture Sensitive Appearance: White Odour: Odourless Melting Point: 2572° Boiling Point: 2850° Solubility: Soluble in water with generation of heat, and forming Ca(OH)2, in acids, glycerol. Insoluble in alcohol Merck Reference: 14,1686 MDL No.: MFCD00010911
Manufacturing of steel, aluminum, magnesium, and glass and calcium salts, building and construction materials, lubricants, laboratory reagent
Formel: CaO
Molecular Weight: 56,08 g/mol
Siedepunkt: 2850 °C
Schmelzpunkt: 2572 °C
Dichte: 3,37
Lagertemperatur: Raumtemperatur
MDL: MFCD00010911
CAS-Nummer: 1305-78-8
EINECS: 215-138-9
UN: 1910
ADR: 8,III
Merck Index: 14,1686