Bariumsulfat ≥99,998% (Metall-Basis), Puratronic®

Lieferant: Thermo Scientific

10651.06 10651.14 10651.22
10651.06EA 43.4 EUR
10651.06 10651.14 10651.22
Bariumsulfat ≥99,998% (Metall-Basis), Puratronic®
Barium sulphate

Purity: 99.998% CAS No.: 7727-43-7 Molecular Formula: BaO4S Formula Weight: 233.40 Physical Form: Powder Density: 4.5 Appearance: White Odour: Odourless Melting Point: 1580° dec. Merck Reference: 14,994 MDL No.: MFCD00003455

Formel: BaSO₄
Molecular Weight: 233,39 g/mol
Siedepunkt: 1600 °C (1013 hPa)
Schmelzpunkt: 1580 °C (Zers.)
Dichte: 4,5
Lagertemperatur: Raumtemperatur
MDL: MFCD00003455
CAS-Nummer: 7727-43-7
EINECS: 231-784-4
Merck Index: 14,994

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR