Strontiumsulfat ≥99,9965% (Metall-Basis), Puratronic®

Lieferant: Thermo Scientific

12870.06 12870.14
12870.06EA 92.2 EUR
12870.06 12870.14
Strontiumsulfat ≥99,9965% (Metall-Basis), Puratronic®
Strontium sulphate

Purity: 99.9965% CAS No.: 7759-02-6 Molecular Formula: O4SSr Formula Weight: 183.68 Physical Form: Powder Density: 3.960 Appearance: White Odour: Odourless Melting Point: 1600° Merck Reference: 14,8848 MDL No.: MFCD00036274

Formel: O₄SSr
Molecular Weight: 183,68 g/mol
Schmelzpunkt: 1600 °C
Dichte: 3,960
Lagertemperatur: Raumtemperatur
MDL: MFCD00036274
CAS-Nummer: 7759-02-6
EINECS: 231-850-2
Merck Index: 14,8848

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR