Strontiumsulfat, Reagenzqualität
Lieferant: Thermo Scientific
38373.14
38373.22
38373.14EA
33
EUR
38373.14
38373.22
Strontiumsulfat, Reagenzqualität
Strontium sulphate
CAS No.: 7759-02-6
Molecular Formula: SrSO4
Formula Weight: 183.68
Physical Form: Crystalline
Density: 3.960
Odour: Odourless
Merck Reference: 14,8848
MDL No.: MFCD00036274
Formel:
O₄SSr Molecular Weight: 183,68 g/mol Schmelzpunkt: 1600 °C Dichte: 3,960 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036274 CAS-Nummer: 7759-02-6 EINECS: 231-850-2 Merck Index: 14,8848 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...