Gallium(II)chlorid, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

413089-1G 413089-5G
SIAL413089-1GEA 141 EUR
SIAL413089-1G SIAL413089-5G
Gallium(II)chlorid, Sigma-Aldrich®
Gallium(II)chlorid
Formel: Ga₂Cl₄
Molecular Weight: 281,26 g/mol
MDL: MFCD00151526
CAS-Nummer: 13498-12-9
UN: 3260
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR