Aluminiumoxid, α-Phase ≥99,9% (Metall-Basis)
Lieferant: Thermo Scientific
Synonyme:
α-Tonerde
42571.22
42571.36
42571.A3
42571.22EA
31.7
EUR
42571.22
42571.36
42571.A3
Aluminiumoxid, α-Phase ≥99,9% (Metall-Basis)
Aluminiumoxid, α-Phase
Purity: 99.9%
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: 20-50 Micron APS, Spherical Powder, S.A. 5-6m2/g
Refractive Index: 1.765
Odour: Odourless
Melting Point: 2045°
Boiling Point: 2980°
Merck Reference: 14,356
MDL No.: MFCD00003424
Formel:
Al₂O₃ Molecular Weight: 101,96 g/mol Siedepunkt: 2980 °C Schmelzpunkt: 2045 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003424 CAS-Nummer: 1344-28-1 EINECS: 215-691-6 Merck Index: 14,356 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...