Indium(III)oxid ≥99,99% (Metall-Basis)
Lieferant: Thermo Scientific
Synonyme:
Diindiumtrioxid, Indiumtrioxid
12544.09
12544.18
12544.30
12544.09EA
117
EUR
12544.09
12544.18
12544.30
Indium(III)oxid ≥99,99% (Metall-Basis)
Indium(III)oxid
Purity: 99.99%
CAS No.: 1312-43-2
Molecular Formula: In2O3
Formula Weight: 277.64
Physical Form: -325 Mesh Powder
Density: 7.18
Melting Point: 850° volat.
Merck Reference: 14,4952
MDL No.: MFCD00011060
Formel:
In₂O₃ Molecular Weight: 277,64 g/mol Schmelzpunkt: 850 °C Dichte: 7,18 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011060 CAS-Nummer: 1312-43-2 EINECS: 215-193-9 Merck Index: 14,4952 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...