Indium(III)oxid ≥99,995% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
Synonyme:
Diindiumtrioxid, Indiumtrioxid
45460.18
45460.09
45460.18EA
439
EUR
45460.18
45460.09
Indium(III)oxid ≥99,995% (Metall-Basis)
Indium(III)oxid
Purity: 99.995%
CAS No.: 1312-43-2
Molecular Formula: In2O3
Formula Weight: 277.64
Physical Form: 6mm & down pieces
Density: 7.18
Melting Point: 850° volat.
Merck Reference: 14,4952
MDL No.: MFCD00011060
Formel:
In₂O₃ Molecular Weight: 277.64 g/mol Schmelzpunkt: 850 °C Dichte: 7.18 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011060 CAS-Nummer: 1312-43-2 EINECS: 215-193-9 Merck Index: 14,4952 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...