Aluminiumhydroxid (hydrargillite), Pulver, EMPLURA®, Supelco®
Lieferant: Merck
Synonyme:
wasserhaltiges Aluminiumoxid
M-Clarity™ Qualitätsstufe = MQ200
1.01091.1000
1.01091.9050
1.01091.1000EA
41.3
EUR
1.01091.1000
1.01091.9050
Aluminiumhydroxid (hydrargillite), Pulver, EMPLURA®, Supelco®
Aluminiumhydroxid
Formel:
Al(OH)₃ Molecular Weight: 78 g/mol Dichte: 2,42 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003420 CAS-Nummer: 21645-51-2 EINECS: 244-492-7 Merck Index: 13,00342 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...