Indium(III)sulfid ≥99,98% (Metall-Basis)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Diindiumtrisulfid, Indiumtrisulfid
44835.04
44835.09
44835.18
44835.04EA
45.1
EUR
44835.04
44835.09
44835.18
Indium(III)sulfid ≥99,98% (Metall-Basis)
Indium(III)sulfid
Purity: 99.98%
CAS No.: 12030-24-9
Molecular Formula: In2S3
Formula Weight: 325.83
Physical Form: Powder
Sensitivity: Moisture Sensitive
MDL No.: MFCD00011061
Formel:
In₂S₃ Molecular Weight: 325,84 g/mol Schmelzpunkt: 1050 °C Dichte: 4,89 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011061 CAS-Nummer: 12030-24-9 EINECS: 234-742-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...