Siliciumcarbid ≥99% (Metall-Basis), Whisker

Lieferant: Thermo Fisher Scientific
Achtung

38787.09 38787.18 38787.30
38787.09EA 80.3 EUR
38787.09 38787.18 38787.30
Siliciumcarbid ≥99% (Metall-Basis), Whisker
Siliciumcarbid

Purity: 99% CAS No.: 409-21-2 Molecular Formula: SiC Formula Weight: 40.10 Physical Form: 1.5 micron dia x approx 18 micron length Density: 3.2 Melting Point: 2700°C Merck Reference: 14,8492 MDL No.: MFCD00049531


Abrasive, polishing, grinding

Formel: SiC
Molecular Weight: 40.1 g/mol
Schmelzpunkt: 2700 °C
Dichte: 3.2
Lagertemperatur: Raumtemperatur
MDL: MFCD00049531
CAS-Nummer: 409-21-2
EINECS: 206-991-8
Merck Index: 14,8492
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd532b391b9c28edcb8adac5fe5ed958ccaa487&vid=d39ce1cfd6c1a5ca44c7d11b656758cbfaed190d&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR