Siliciumcarbid, Pulver, superfein 600 grit

Lieferant: Thermo Fisher Scientific
Achtung

A13561.0C A13561.0E A13561.36
A13561.0CEA 536 EUR
A13561.0C A13561.0E A13561.36
Siliciumcarbid, Pulver, superfein 600 grit
Siliciumcarbid

CAS No.: 409-21-2 Molecular Formula: SiC Formula Weight: 40.10 Odour: Odourless MDL No.: MFCD00049531

Formel: SiC
Molecular Weight: 40.1 g/mol
Schmelzpunkt: 2700
Dichte: 3.16
Lagertemperatur: Raumtemperatur
MDL: MFCD00049531
CAS-Nummer: 409-21-2
EINECS: 206-991-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd532b391b9c28edcb8adac5fe5ed958ccaa487&vid=d39ce1cfd6c1a5ca44c7d11b656758cbfaed190d&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR