Blei(II)oxid ≥99,999% (Metall-Basis), Puratronic®

Lieferant: Thermo Scientific
Gefahr

Synonyme: Bleimonoxid

40753.14 40753.22 40753.36
40753.14EA 86.9 EUR
40753.14 40753.22 40753.36
Blei(II)oxid ≥99,999% (Metall-Basis), Puratronic®
Blei(II)oxid

Purity: 99.999% CAS No.: 1317-36-8 Molecular Formula: PbO Formula Weight: 223.19 Physical Form: Powder Density: 9.53 Appearance: Yellow Odour: Odourless Melting Point: 886° Merck Reference: 14,5413 UN No.: 3077 MDL No.: MFCD00011164

Formel: PbO
Molecular Weight: 223,2 g/mol
Siedepunkt: 1470 °C (1013 hPa)
Schmelzpunkt: 886 °C
Dichte: 9,53
Lagertemperatur: Raumtemperatur
MDL: MFCD00011164
CAS-Nummer: 1317-36-8
EINECS: 215-267-0
UN: 3077
ADR: 9,III
Merck Index: 14,5413

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR