Blei(II)fluorid ≥99,997% (Metall-Basis), Puratronic®
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Lead difluoride
10723.14
10723.22
10723.14EA
119
EUR
10723.14
10723.22
Blei(II)fluorid ≥99,997% (Metall-Basis), Puratronic®
Blei(II)fluorid
Purity: 99.997%
CAS No.: 7783-46-2
Molecular Formula: PbF2
Formula Weight: 245.20
Physical Form: Powder
Density: 8.445
Melting Point: 855°
Boiling Point: 1290°
Merck Reference: 14,5408
UN No.: 2291
MDL No.: MFCD00011162
Formel:
F₂Pb Molecular Weight: 245,2 g/mol Siedepunkt: 1290 °C Schmelzpunkt: 855 °C Dichte: 8,445 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011162 CAS-Nummer: 7783-46-2 EINECS: 231-998-8 UN: 2291 ADR: 6.1,III Merck Index: 14,5408 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...