Blei(II)iodid, ultra trocken ≥99,999% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Bleidiiodid

44314.06 44314.14 44314.22
44314.06EA 112 EUR
44314.06 44314.14 44314.22
Blei(II)iodid, ultra trocken ≥99,999% (Metall-Basis)
Blei(II)iodid

Purity: 99.999% CAS No.: 10101-63-0 Molecular Formula: I2Pb Formula Weight: 461.00 Physical Form: -10 Mesh Beads Density: 6.16 Appearance: Yellow Odour: Odourless Melting Point: 402° Boiling Point: 954° Merck Reference: 14,5411 UN No.: 2291 MDL No.: MFCD00011163

Formel: I₂Pb
Molecular Weight: 461,01 g/mol
Siedepunkt: 954 °C
Schmelzpunkt: 402 °C
Dichte: 6,16
Lagertemperatur: Raumtemperatur
MDL: MFCD00011163
CAS-Nummer: 10101-63-0
EINECS: 233-256-9
UN: 2291
ADR: 6.1,III
Merck Index: 14,5411

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR