Germanium(II)iodid ≥99,99% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Germaniumdiiodid

14013.03 14013.06
14013.03EA 219 EUR
14013.03 14013.06
Germanium(II)iodid ≥99,99% (Metall-Basis)
Germanium(II)iodid

Purity: 99.99% CAS No.: 13573-08-5 Molecular Formula: GeI2 Formula Weight: 326.40 Physical Form: -10 Mesh Density: 5.37 Appearance: Yellow to brown UN No.: 3260 MDL No.: MFCD00135537

Formel: GeI₂
Molecular Weight: 326.42 g/mol
Schmelzpunkt: 460
Dichte: 5.37
Lagertemperatur: Raumtemperatur
MDL: MFCD00135537
CAS-Nummer: 13573-08-5
EINECS: 236-998-1
UN: 3260
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR