Zinn(IV)iodid ≥95%

Lieferant: Thermo Fisher Scientific
Gefahr

71114.09 71114.18
71114.09EA 71.1 EUR
71114.09 71114.18
Zinn(IV)iodid ≥95%
Zinn(IV)iodid

Purity: 95% CAS No.: 7790-47-8 Molecular Formula: I4Sn Formula Weight: 626.31 Physical Form: Crystalline Density: 4.46 Refractive Index: 2.106 Sensitivity: Moisture Sensitive Appearance: Red Melting Point: ca 143° Boiling Point: 340° Solubility: Soluble in alcohol, benzene, CS2, chloroform, and ether. Decomposes in water Merck Reference: 14,8777 UN No.: 3260 MDL No.: MFCD00036272

Formel: I₄Sn
Siedepunkt: 340 °C
Schmelzpunkt: ∼ 143 °C
Dichte: 4,46
Lagertemperatur: Raumtemperatur
MDL: MFCD00036272
CAS-Nummer: 7790-47-8
EINECS: 232-208-4
UN: 3260
ADR: 8,III
Merck Index: 14,8777

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR