Kieselgur -325 Mesh Powder
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: Diatomaceous silica, Diatomaceous earth, Siliceous earth, Infusorial earth, Diatomite, Celite
89381.A1 89381.A9
89381.A1EA 66 EUR
89381.A1 89381.A9
Kieselgur -325 Mesh Powder
Kieselgur
Kieselguhr is used in used as a filtration aid, mild abrasive in products including metal polishes and toothpaste, absorbent for liquids, matting agent for coatings, reinforcing filler in plastics and rubber, anti-block in plastic films, porous support for chemical catalysts, cat litter, activator in blood clotting studies, a stabilizing component of dynamite, and a thermal insulator.
CAS No.: 7631-86-9 Physical Form: -325 Mesh Powder Density: 2.0 Appearance: Off-white to cream Odour: Odourless Melting Point: 1300° dec. MDL No.: MFCD00132803
Formel: O₂Si
Molecular Weight: 60.08 g/mol
Siedepunkt: 2230
Schmelzpunkt: >1300 °C (Zers.)
Dichte: 2.0
Lagertemperatur: Raumtemperatur
MDL: MFCD00132803
CAS-Nummer: 7631-86-9
EINECS: 231-545-4