Siliciumdioxid ≥99,5% (Metall-Basis)
Lieferant: Thermo Scientific
Gefahr
Synonyme: Silicium(IV)oxid
88316.30 88316.A1
88316.30EA 106 EUR
88316.30 88316.A1
Siliciumdioxid ≥99,5% (Metall-Basis)
Siliciumdioxid
Silicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.
Purity: 99.5% CAS No.: 14808-60-7 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: -325 Mesh Powder Density: 2.2-2.6 Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232
Formel: SiO₂
Molecular Weight: 60,08 g/mol
Siedepunkt: 2230 °C
Schmelzpunkt: 1710 °C
Dichte: 2,2…2.6
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 14808-60-7
EINECS: 238-878-4
Merck Index: 14,8493