Germanium(IV)isopropoxid ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Germaniumtetraisopropoxid, Germanium(IV)isopropylat, Tetraisopropylorthogermanat, Germaniumtetraisopropanolat, Germaniumtetraisopropylat, Germanium(IV)isopropanolat

42673.03 42673.06 42673.14
42673.03EA 69.1 EUR
42673.03 42673.06 42673.14
Germanium(IV)isopropoxid ≥99,9% (Metall-Basis)
Germanium(IV)isopropoxid

Purity: 99.9% CAS No.: 21154-48-3 Molecular Formula: C12H28GeO4 Formula Weight: 308.94 Flash Point: 68°(154°F) Physical Form: Liquid Density: 1.025 Refractive Index: 1.414 Sensitivity: Moisture Sensitive Appearance: Colourless Odour: Alcohol-like Boiling Point: 109°/30mm MDL No.: MFCD00015339

Formel: C₁₂H₂₈GeO₄
Molecular Weight: 308.96 g/mol
Siedepunkt: 109 °C (30 mmHg)
Dichte: 1.025
Flammpunkt: 68 °C (154 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00015339
CAS-Nummer: 21154-48-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR