Bleiselenid ≥99,999% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Lead(II) selenide

44916.22 44916.14
44916.22EA 953 EUR
44916.22 44916.14
Bleiselenid ≥99,999% (Metall-Basis)
Bleiselenid

Purity: 99.999+% CAS No.: 12069-00-0 Molecular Formula: PbSe Formula Weight: 286.15 Physical Form: 3-6mm Fused Lump Odour: Product specific UN No.: 3077 MDL No.: MFCD00016273

Formel: PbSe
Molecular Weight: 286,16 g/mol
Schmelzpunkt: 79,5…80 °C
Dichte: 8,1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00016273
CAS-Nummer: 12069-00-0
EINECS: 235-109-4
UN: 3077
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR