Zinn(II)sulfat ≥95,5%
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
Zinnsulfat
11537.30
11537.A1
11537.30EA
75.1
EUR
11537.30
11537.A1
Zinn(II)sulfat ≥95,5%
Zinn(II)sulfat
Purity: 95.5% min
CAS No.: 7488-55-3
Molecular Formula: SnSO4
Formula Weight: 214.77
Physical Form: Powder
Density: 4.15
Melting Point: 378° dec.
Solubility: Soluble in water and dilute H2SO4. Water solution hydrolyzes rapidly with precipitation of basic salt
Merck Reference: 14,8790
MDL No.: MFCD00011246
Tin plating baths
Formel:
O₄SSn Molecular Weight: 214,77 g/mol Schmelzpunkt: 378 °C (Zers.) Dichte: 4,15 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011246 CAS-Nummer: 7488-55-3 EINECS: 231-302-2 Merck Index: 14,8790 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...