Vinyltriethoxysilan ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Triethoxy(vinyl)silan
B21037.0B
B21037.18
B21037.30
B21037.0BEA
171
EUR
B21037.0B
B21037.18
B21037.30
Vinyltriethoxysilan ≥97%
Vinyltriethoxysilan
Purity: 97%
CAS No.: 78-08-0
Molecular Formula: C8H18O3Si
Formula Weight: 190.31
Flash Point: 34°(93°F)
Physical Form: Liquid
Density: 0.903
Refractive Index: 1.3990
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Fruit-like
Boiling Point: 160-161°
Beilstein Reference: 1767229
UN No.: 1993
MDL No.: MFCD00009063
Formel:
C₈H₁₈O₃Si Molecular Weight: 190,31 g/mol Siedepunkt: 160…161 °C Dichte: 0,903 Flammpunkt: 34 °C (93 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009063 CAS-Nummer: 78-08-0 EINECS: 201-081-7 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...