(3-Brompropyl)trimethoxysilan

Lieferant: CHEMPUR SILANE

Synonyme: Trimethoxy(3-bromopropyl)silane, 3-(Trimethoxysilyl)propylbromide

SILS03125.10 SILS03125.25 SILS03125.100
SILNSILS03125.10EA 0 EUR
SILNSILS03125.10 SILNSILS03125.25 SILNSILS03125.100
(3-Brompropyl)trimethoxysilan
(3-Brompropyl)trimethoxysilan
Formel: BrH₂CCH₂CH₂Si(OCH₃)₃
Molecular Weight: 243,17 g/mol
MDL: MFCD00053894
CAS-Nummer: 51826-90-5
EINECS: 257-453-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR