(3-Glycidoxypropyl)triethoxysilan
Lieferant: CHEMPUR SILANE
Synonyme:
(3-Glycidyloxypropyl)triethoxysilan, Geniosil GF 82, (3-(2,3-Epoxypropoxy)propyl)triethoxysilan
SILS09135.25
SILS09135.100
SILNSILS09135.25EA
0
EUR
SILNSILS09135.25
SILNSILS09135.100
(3-Glycidoxypropyl)triethoxysilan
(3-Glycidoxypropyl)triethoxysilan
Formel:
C₁₂H₂₆O₅Si Molecular Weight: 278,42 g/mol |
MDL:
MFCD01311705 CAS-Nummer: 2602-34-8 EINECS: 220-011-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...