(3-Glycidoxypropyl)trimethoxysilan ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (3-Glycidyloxypropyl)trimethoxysilan, (3-(2,3-Epoxypropoxy)propyl)trimethoxysilan

A18431.14 A18431.36 A18431.22 AB116946.0025
A18431.14EA 35.1 EUR
A18431.14 A18431.36 A18431.22 ABCRAB116946.0025
(3-Glycidoxypropyl)trimethoxysilan ≥97%
(3-Glycidoxypropyl)trimethoxysilan

Purity: 97% CAS No.: 2530-83-8 Molecular Formula: C9H20O5Si Formula Weight: 236.34 Flash Point: 135°(275°F) Refractive Index: 1.4290 Odour: Odourless Melting Point: -70° Boiling Point: 119-121°/2mm Beilstein Reference: 4308125 MDL No.: MFCD00005144

Formel: C₉H₂₀O₅Si
Molecular Weight: 236,34 g/mol
Siedepunkt: 119…121 °C (2 mmHg)
Schmelzpunkt: < –70 °C
Dichte: 1,072
Flammpunkt: 135 °C (275 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005144
CAS-Nummer: 2530-83-8
EINECS: 219-784-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR