Methyltrichlorsilan ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Trichlor(methyl)silan, Methylsilyltrichlorid

B23107.22 B23107.36
B23107.22EA 33.4 EUR
B23107.22 B23107.36
Methyltrichlorsilan ≥97%
Methyltrichlorsilan

Purity: 97% CAS No.: 75-79-6 Molecular Formula: CH3Cl3Si Formula Weight: 149.48 Flash Point: 8°(46°F) Refractive Index: 1.4115 Odour: Pungent Melting Point: -78° Boiling Point: 64-66° Beilstein Reference: 1361381 UN No.: 1250 MDL No.: MFCD00000481

Formel: CH₃Cl₃Si
Molecular Weight: 149,48 g/mol
Siedepunkt: 64…66 °C
Schmelzpunkt: –78 °C
Dichte: 1,270
Flammpunkt: 8 °C (46 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000481
CAS-Nummer: 75-79-6
EINECS: 200-902-6
UN: 1250
ADR: 3,II
REACH: 01-2119475226-36

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR