(3-Chlorpropyl)triethoxysilan

Lieferant: CHEMPUR SILANE

Synonyme: 3-(Triethoxysilyl)propylchlorid, Triethoxy(3-chlorpropyl)silan

SILS04650.25 SILS04650.100
SILNSILS04650.25EA 0 EUR
SILNSILS04650.25 SILNSILS04650.100
(3-Chlorpropyl)triethoxysilan
(3-Chlorpropyl)triethoxysilan
Formel: ClH₂CCH₂CH₂Si(OCH₂CH₃)₃
Molecular Weight: 240,8 g/mol
MDL: MFCD00018985
CAS-Nummer: 5089-70-3
EINECS: 225-805-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR