(3-Chlorpropyl)trimethoxysilan ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Trimethoxy(3-chlorpropyl)silan, 3-(Trimethoxysilyl)propylchlorid

verpackt unter Argon in wiederversiegelbaren ChemSeal™ Flaschen

42413.AD 42413.AK 42413.K2
42413.ADEA 29.1 EUR
42413.AD 42413.AK 42413.K2
(3-Chlorpropyl)trimethoxysilan ≥97%
(3-Chlorpropyl)trimethoxysilan

Packaged under Argon in resealable ChemSeal® bottles


Purity: 97+% CAS No.: 2530-87-2 Molecular Formula: ClCH2CH2CH2Si(OCH3)3 Formula Weight: 198.72 Flash Point: 78°(172°F) Physical Form: Liquid Density: 1.077 Refractive Index: 1.4200 Sensitivity: Moisture Sensitive Odour: Recognizable Melting Point: -50° Boiling Point: 195-197° Beilstein Reference: 969627 MDL No.: MFCD00000997

Formel: ClH₂CCH₂CH₂Si(OCH₃)₃
Molecular Weight: 198,72 g/mol
Siedepunkt: 195…197 °C
Schmelzpunkt: –50 °C
Dichte: 1,077
Flammpunkt: 78 °C (172 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000997
CAS-Nummer: 2530-87-2
EINECS: 219-787-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR