(2-Cyanethyl)triethoxysilan ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Triethoxy(2-cyanethyl)silan, 3-(Triethoxysilyl)propionitril, 3-(Triethoxysilyl)propiononitril

L03543.14 L03543.22
L03543.14EA 47.1 EUR
L03543.14 L03543.22
(2-Cyanethyl)triethoxysilan ≥97%
(2-Cyanethyl)triethoxysilan

Purity: 97% CAS No.: 919-31-3 Molecular Formula: NCCH2CH2Si(OCH2CH3)3 Formula Weight: 217.34 Flash Point: 100°(212°F) Refractive Index: 1.4130 Boiling Point: 224-225° Beilstein Reference: 1776392 MDL No.: MFCD00019851

Formel: NCCH₂CH₂Si(OCH₂CH₃)₃
Molecular Weight: 217,34 g/mol
Siedepunkt: 224…225 °C
Dichte: 0,980
Flammpunkt: 100 °C (212 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00019851
CAS-Nummer: 919-31-3
EINECS: 213-050-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR