(2-Cyanethyl)triethoxysilan

Lieferant: CHEMPUR SILANE

Synonyme: Triethoxy(2-cyanethyl)silan, 3-(Triethoxysilyl)propionitril, 3-(Triethoxysilyl)propiononitril

SILS04875.10 SILS04875.25 SILS04875.100
SILNSILS04875.10EA 0 EUR
SILNSILS04875.10 SILNSILS04875.25 SILNSILS04875.100
(2-Cyanethyl)triethoxysilan
(2-Cyanethyl)triethoxysilan
Formel: NCCH₂CH₂Si(OCH₂CH₃)₃
Molecular Weight: 217,34 g/mol
Siedepunkt: 133 °C (28 mmHg)
Dichte: 0,9792 g/cm³ (20 °C)
MDL: MFCD00019851
CAS-Nummer: 919-31-3
EINECS: 213-050-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR