(2-Cyanethyl)trimethoxysilan

Lieferant: CHEMPUR SILANE

SILS04900.5 SILS04900.25 SILS04900.100
SILNSILS04900.5EA 0 EUR
SILNSILS04900.5 SILNSILS04900.25 SILNSILS04900.100
(2-Cyanethyl)trimethoxysilan
(2-Cyanethyl)trimethoxysilan
Formel: NCCH₂CH₂Si(OCH₃)₃
MDL: MFCD00039494
CAS-Nummer: 2526-62-7
EINECS: 219-764-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR