(3-(Diethylamino)propyl)trimethoxysilan
Lieferant: CHEMPUR SILANE
Synonyme:
N,N-Diethyl-3-(trimethoxysilyl)propylamin, Trimethoxy(3-(diethylamino)propyl)silan, (3-Diethylaminopropyl)trimethoxysilan
SILS06300.5
SILS06300.25
SILS06300.100
SILNSILS06300.5EA
0
EUR
SILNSILS06300.5
SILNSILS06300.25
SILNSILS06300.100
(3-(Diethylamino)propyl)trimethoxysilan
(3-(Diethylamino)propyl)trimethoxysilan
Formel:
(H₃CCH₂)₂NCH₂CH₂CH₂Si(OCH₃)₃ Molecular Weight: 235,4 g/mol |
MDL:
MFCD00039882 CAS-Nummer: 41051-80-3 EINECS: 255-192-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...