(3-(Dimethylamino)propyl)trimethoxysilan
Lieferant: CHEMPUR SILANE
Synonyme:
Trimethoxy(3-(dimethylamino)propyl)silan, N,N-Dimethyl-3-(trimethoxysilyl)propylamin, (3-Dimethylaminopropyl)trimethoxysilan
SILS06900.5
SILS06900.25
SILNSILS06900.5EA
0
EUR
SILNSILS06900.5
SILNSILS06900.25
(3-(Dimethylamino)propyl)trimethoxysilan
(3-(Dimethylamino)propyl)trimethoxysilan
Formel:
(H₃C)₂NCH₂CH₂CH₂Si(OCH₃)₃ Molecular Weight: 207.35 g/mol |
MDL:
MFCD00053674 CAS-Nummer: 2530-86-1 EINECS: 219-786-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...