(3-Aminopropyl)trimethoxysilan 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Trimethoxy(3-aminopropyl)silan, 3-(Trimethoxysilyl)propylamin

A11284.0E A11284.22 A11284.36
A11284.0EEA 775 EUR
A11284.0E A11284.22 A11284.36
(3-Aminopropyl)trimethoxysilan 97%
(3-Aminopropyl)trimethoxysilan

Purity: 97% CAS No.: 13822-56-5 Molecular Formula: C6H17NO3Si Formula Weight: 179.29 Flash Point: 90°(194°F) Physical Form: Liquid Density: 1.02 Refractive Index: 1.4240 Sensitivity: Air and Moisture Sensitive Appearance: Colourless Odour: Amine-like Boiling Point: 194° Beilstein Reference: 2038118 UN No.: 2735 MDL No.: MFCD00008206

Formel: H₂NCH₂CH₂CH₂Si(OCH₃)₃
Molecular Weight: 179.29 g/mol
Siedepunkt: 194 °C
Dichte: 1.02
Flammpunkt: 90 °C (194 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008206
CAS-Nummer: 13822-56-5
EINECS: 237-511-5
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR