(3-(Methylamino)propyl)trimethoxysilan

Lieferant: abcr

Synonyme: Trimethoxy(3-(methylamino)propyl)silan, N-Methyl-3-(trimethoxysilyl)propylamin, (3-Methylaminopropyl)trimethoxysilan

AB111230.0025 AB111230.0100
ABCRAB111230.0025EA 0 EUR
ABCRAB111230.0025 ABCRAB111230.0100
(3-(Methylamino)propyl)trimethoxysilan
(3-(Methylamino)propyl)trimethoxysilan
Formel: H₃CNHCH₂CH₂CH₂Si(OCH₃)₃
Molecular Weight: 193.32 g/mol
MDL: MFCD00048021
CAS-Nummer: 3069-25-8
EINECS: 221-334-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR