(3-(Methylamino)propyl)trimethoxysilan
Lieferant: CHEMPUR SILANE
Synonyme:
Trimethoxy(3-(methylamino)propyl)silan, N-Methyl-3-(trimethoxysilyl)propylamin, (3-Methylaminopropyl)trimethoxysilan
SILS11025.25
SILS11025.100
SILNSILS11025.25EA
0
EUR
SILNSILS11025.25
SILNSILS11025.100
(3-(Methylamino)propyl)trimethoxysilan
(3-(Methylamino)propyl)trimethoxysilan
Formel:
H₃CNHCH₂CH₂CH₂Si(OCH₃)₃ Molecular Weight: 193,32 g/mol |
MDL:
MFCD00048021 CAS-Nummer: 3069-25-8 EINECS: 221-334-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...